£0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
United Kingdom
ContactUsLinkComponent
(2,4-dichloro-5,7-dihydropyrrolo[3,4-d]pyrimidin-6-yl)-(4-methoxytetrahydropyran-4-yl)methanone 1 * 1 g
PHBLPBPW019-1G
:
Pharmablock (USA)
(2,4-dichloro-5,7-dihydropyrrolo[3,4-d]pyrimidin-6-yl)-(4-methoxytetrahydropyran-4-yl)methanone 1 * 1 g
PHBLPBPW019-1G
:
Pharmablock (USA)
Frequently Bought Together
Isoamyl alcohol, TECHNICAL
: VWR Chemicals
ACETONITRILE EMPLURA® 1 * 4 L
: Merck
Methyl acetate anhydrous, 99.5 % 1 * 1 L
: Merck
(±)-2-Methyltetrahydrofuran 99+% stabilised, pure
: Thermo Fisher Scientific
Butyl acetate 99+% for spectroscopy
: Thermo Fisher Scientific
Petroleum spirit, 60…80 °C
: Thermo Fisher Scientific
Chloroform ≥99.5% stabilised
: Honeywell Chemicals
2-Propanol ≥99.9%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
: Honeywell Chemicals
1,2-Dimethoxyethane, extra dry 99+% stabilised, AcroSeal®
: Thermo Fisher Scientific
Ethyl acetate ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
: VWR Chemicals
Cyclohexane 99.8% for HPLC
: Thermo Fisher Scientific
Dimethyl sulphoxide 99.9+% ACS
: Thermo Fisher Scientific
1-Butanol 99%, extra pure
: Thermo Fisher Scientific