2-chloro-1-(oxan-2-yl)-5-(tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-imidazole 1 * 1 g
Catalog # PHBLPBN20121887-1G
Supplier: Pharmablock (USA)
2-chloro-1-(oxan-2-yl)-5-(tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-imidazole 1 * 1 g
Catalog # PHBLPBN20121887-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Petroleum spirit, 40…60 °C, GPR RECTAPUR®
Supplier: VWR Chemicals
1-Propanol, EMPLURA®, Supelco®
Supplier: Merck
Tetrahydrofuran ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Toluene ≥99.8%, SupraSolv® for GC-MS
Supplier: Merck
Ethanol 96% ≥96% (v/v), TechniSolv®, pure
Supplier: VWR Chemicals
tert-Butyl methyl ether, Uvasol® for spectroscopy, Supelco®
Supplier: Merck
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Dichloromethane ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
Xylene (mixture of isomers) ≥98%, TECHNICAL
Supplier: VWR Chemicals
Ethanol, GPR RECTAPUR®
Supplier: VWR Chemicals
Glycerine ≥99%
Supplier: Thermo Fisher Scientific
Toluene ≥99.8% (by GC), ChromAR® for liquid chromatography, for UV spectrophotometry, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND