ethyl 5,6,7,8-tetrahydro-1,7-naphthyridine-3-carboxylate hydrochloride 1 * 1 g
Catalog # PHBLPBN20114071-1G
Supplier: Pharmablock (USA)
ethyl 5,6,7,8-tetrahydro-1,7-naphthyridine-3-carboxylate hydrochloride 1 * 1 g
Catalog # PHBLPBN20114071-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Ethanol 20% (w/w), Emprove® Expert for cleaning biochromatography resins, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Dichloromethane, dehydrated (max. 0.01% H₂O) ≥99% stabilised, GPR RECTAPUR® for synthesis
Supplier: VWR Chemicals
Methyl ethyl ketone ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Methanol ≥99.9%, SPECTRONORM® for spectroscopy
Supplier: VWR Chemicals
Acetone, for GC Plus, Burdick & Jackson™
Supplier: Honeywell Chemicals
Isoamyl Alcohol according to Gerber for analysis 1 * 1.000 mL
Supplier: PanReac AppliChem
Ethyl acetate, AR®, Laboratory Reagent, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
Cesium chloride 99.99% (trace metals basis), Optical Grade for spectroscopy
Supplier: Thermo Fisher Scientific
N-Methyl-2-pyrrolidone (NMP) 99+% ACS
Supplier: Thermo Fisher Scientific
Hydrindantin dihydrate, Supelco®
Supplier: Merck
Chloroform ≥99.8% stabilised ACS, meets analytical specifications of Reagent Ph. Eur.
Supplier: Thermo Fisher Scientific
Tetrahydrofuran ≥99.7% unstabilised for HPLC, for UV spectrophotometry
Supplier: Thermo Fisher Scientific
Acetonitrile, extra dry over Molecular Sieve 99.9%, AcroSeal®
Supplier: Thermo Fisher Scientific
Tetrahexylammonium bromide, Supelco®
Supplier: Merck
Cyclohexane 99+% ACS, meets analytical specifications of Reagent Ph. Eur. for spectroscopy
Supplier: Thermo Fisher Scientific
Tetrabutylammonium dihydrogen phosphate 99% for HPLC
Supplier: Thermo Fisher Scientific
ETHYL ACETATE EXTRA PURE, DAC, N. F. 1 * 100 L
Supplier: Honeywell Chemicals
Chloroform ≥99.5% stabilised
Supplier: Honeywell Chemicals