(3aS,5R,6S,6aS)-5-{[(tert-butyldiphenylsilyl)oxy]methyl}-2,2-dimethyl-tetrahydro-2H-furo[2,3-d][1,3]dioxol-6-ol 1 * 1 g
Catalog # PHBLPBLY8053-1G
Supplier: Pharmablock (USA)
(3aS,5R,6S,6aS)-5-{[(tert-butyldiphenylsilyl)oxy]methyl}-2,2-dimethyl-tetrahydro-2H-furo[2,3-d][1,3]dioxol-6-ol 1 * 1 g
Catalog # PHBLPBLY8053-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Water, GPR RECTAPUR®
Supplier: VWR Chemicals
2-Propanol ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Petroleum spirit, 40…60 °C, GPR RECTAPUR®
Supplier: VWR Chemicals
1-Propanol, LiChrosolv® for HPLC, Supelco®
Supplier: Merck
Dichloromethane, dehydrated (max. 0.01% H₂O) ≥99% stabilised, GPR RECTAPUR® for synthesis
Supplier: VWR Chemicals
Xylene (mixture of isomers) ≥98.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
tert-Butyl methyl ether, Uvasol® for spectroscopy, Supelco®
Supplier: Merck
2-Propanol ≥98%, TECHNICAL
Supplier: VWR Chemicals
Pyridine ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
n-Heptane, LiChrosolv® for liquid chromatography, Supelco®
Supplier: Merck
Diethyl ether ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
(±)-2-Butanol, EMSURE® for analysis, Supelco®
Supplier: Merck
Methyl ethyl ketone ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals