(4aS,9bR)-6-bromo-1H,2H,3H,4H,4aH,5H,9bH-pyrido[4,3-b]indole 1 * 1 g
Catalog # PHBLPBLJ7681-5-1G
Supplier: Pharmablock (USA)
(4aS,9bR)-6-bromo-1H,2H,3H,4H,4aH,5H,9bH-pyrido[4,3-b]indole 1 * 1 g
Catalog # PHBLPBLJ7681-5-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Ethanol (taxed), pure for molecular biology (200 Proof), Riedel-de Haën™
Supplier: Honeywell Chemicals
Dimethyl sulfoxide EMPLURA 1 * 185 L
Supplier: Merck
Water, GPR RECTAPUR®
Supplier: VWR Chemicals
2-Methylbutane 99+%, extra pure
Supplier: Thermo Fisher Scientific
Benzonitrile 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
ETHYL ACETATE 99.5% FOR SYNTHESIS 1 * 200 L
Supplier: PanReac AppliChem
Pentane reagent grade, 98% 1 * 1 L
Supplier: Merck
Dimethyl sulphoxide, extra dry over Molecular Sieve 99.7+%, AcroSeal®
Supplier: Thermo Fisher Scientific
Dimethyl sulphoxide ≥99%
Supplier: Thermo Fisher Scientific
Acetonitrile ≥99.5% ACS
Supplier: Thermo Fisher Scientific
Chloroform ≥99.8% stabilised ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Chloroform ≥99.5% stabilised
Supplier: Honeywell Chemicals