(2S)-2-hydroxy-2-phenylacetic acid (4aS,9bR)-6-bromo-1H,2H,3H,4H,4aH,5H,9bH-pyrido[4,3-b]indole 1 * 1 g
PHBLPBLJ7681-1G
(2S)-2-hydroxy-2-phenylacetic acid (4aS,9bR)-6-bromo-1H,2H,3H,4H,4aH,5H,9bH-pyrido[4,3-b]indole 1 * 1 g
PHBLPBLJ7681-1G
Frequently Bought Together
Dichloromethane ≥99% stabilised, GPR RECTAPUR®
: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
Ethanol 96% vol Ph. Eur.
: VWR Chemicals
Petroleum spirit, 40…60 °C, GPR RECTAPUR®
: VWR Chemicals
Water, AnalaR NORMAPUR® ISO 3696 Grade 3 analytical reagent
: VWR Chemicals
1-Pentanol ≥98%, GPR RECTAPUR®
: VWR Chemicals
Water USP, JP, Ph. Eur., purified
: VWR Chemicals
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals