2,5-dichloro-N-(5-methyl-1H-pyrazol-3-yl)pyrimidin-4-amine 1 * 1 g
Catalog # PHBLPBLJ7191-1G
Supplier: Pharmablock (USA)
2,5-dichloro-N-(5-methyl-1H-pyrazol-3-yl)pyrimidin-4-amine 1 * 1 g
Catalog # PHBLPBLJ7191-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Ethyl acetate ≥99.7%, PESTINORM® for pesticide residue analysis
Supplier: VWR Chemicals
Ethyl acetate ≥99.5% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Ethanol (untaxed) in water ≥70% (v/v) Ph. Helv., Riedel-de Haën™
Supplier: Honeywell Chemicals
Xylene (mixture of isomers) ≥98%, liquid
Supplier: PanReac AppliChem
2,4,6-Tris(2-pyridyl)-1,3,5-triazine, Supelco®
Supplier: Merck
Petroleum spirit, 80…110 °C
Supplier: Roth Carl
Diisopropyl ether 99+% stabilised, extra pure
Supplier: Thermo Fisher Scientific
Dimethyl sulphoxide, extra dry over Molecular Sieve 99.7+%, AcroSeal®
Supplier: Thermo Fisher Scientific
N-Methyl-2-pyrrolidone (NMP) ≥99.0% ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals
n-Hexane 95%, ChromAR®, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
1,2-Dichlorobenzene 99+% for HPLC
Supplier: Thermo Fisher Scientific
Dimethyl sulphoxide (DMSO),?99,9 %, Ph. Eur., USP 1 * 500 mL
Supplier: Roth Carl