tert-butyl N-[cis-3-methylpiperidin-4-yl]carbamate 1 * 1 g
Catalog # PHBLPBLJ3573-1G
Supplier: Pharmablock (USA)
tert-butyl N-[cis-3-methylpiperidin-4-yl]carbamate 1 * 1 g
Catalog # PHBLPBLJ3573-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Tetrachloroethylene, EMPLURA®, Supelco®
Supplier: Merck
1,4-Dioxane stabilised, EMPLURA®, Supelco®
Supplier: Merck
Dichloromethane, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
Dimethyl sulphoxide, EMPLURA®, Supelco®
Supplier: Merck
Methanol ≥99.9% (by GC) Reag. Ph. Eur., ACS, ISO for analysis
Supplier: PanReac AppliChem
m-Xylene, extra dry 99%, AcroSeal®
Supplier: Thermo Fisher Scientific
Tetraheptylammonium bromide, Supelco®
Supplier: Merck
Chlorobenzene ≥99%
Supplier: Thermo Fisher Scientific
1-Butanol ≥99.7%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Tetrahydrofuran 99.8% unstabilised for HPLC
Supplier: Thermo Fisher Scientific
n-Nonane 99%, Reagent Grade
Supplier: Honeywell Chemicals
Methanol ≥99.9%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
2-Propanol ≥99.8%, BAKER ANALYZED® for LC-MS, J.T.Baker®
Supplier: Avantor
Isobutanol, Sigma-Aldrich®
Supplier: Merck
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals