cis-3-fluoro-4-methoxy-pyrrolidine hydrochloride 1 * 1 g
PHBLPBLJ2552-1-1G
Frequently Bought Together
o-Xylene anhydrous, 97 % 1 * 2 L
: Merck
4,4'-Bis(dimethylamino)diphenylmethane 98%
: Thermo Fisher Scientific
n-Pentane, extra dry 99+%, AcroSeal®
: Thermo Fisher Scientific
Water, pure, demineralised
: Thermo Fisher Scientific
1,2-Dichlorobenzene 99+% for spectroscopy
: Thermo Fisher Scientific
Isooctane (Reag. Ph. Eur.) for analysis, ACS 1 * 2,5 L
: PanReac AppliChem
3-(Chloromethyl)benzoyl chloride 98%
: Thermo Fisher Scientific
Ethanolamine, Sigma-Aldrich®
: Merck
ASSAY VOLLSTÄNDIGE ZELLE ABLÖSUNG REAGEN
: DISCOVERX
1-Propanol ≥99%, GPR RECTAPUR®
: VWR Chemicals
Ethylene glycol monomethyl ether (2-methoxyethanol) 99+% for spectroscopy
: Thermo Fisher Scientific
n-Nonane 99%, Reagent Grade
: Honeywell Chemicals
Diisopropyl ether stabilised for analysis
: Thermo Fisher Scientific
Isooctane for UV, IR, HPLC, ACS 1 * 1 L
: PanReac AppliChem