2-chloro-1-(5-methoxy-1H-indol-3-yl)ethan-1-one 1 * 1 g
Catalog # PHBLPBLJ2145-1G
Supplier: Pharmablock (USA)
2-chloro-1-(5-methoxy-1H-indol-3-yl)ethan-1-one 1 * 1 g
Catalog # PHBLPBLJ2145-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
tert-Butyl methyl ether anhydrous, 99.8 % 1 * 1 L
Supplier: Merck
(±)-Tetrahydrofurfuryl alcohol, Sigma-Aldrich®
Supplier: Merck
Toluene 99.5% ACS for spectroscopy
Supplier: Thermo Fisher Scientific
Methanol ≥99.9% (by GC) Reag. Ph. Eur., ACS, ISO for analysis
Supplier: PanReac AppliChem
N,N-Dimethylformamide 99+%, extra pure
Supplier: Thermo Fisher Scientific
Methanol ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
White spirit
Supplier: BARRETTINE INDUSTRIAL
Acetonitrile ≥99.9%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
n-Hexane
Supplier: Roth Carl
Isoamyl alcohol ≥98.5% (by GC), Puriss. p.a. ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Isoamyl Alcohol according to Gerber for analysis 1 * 1.000 mL
Supplier: PanReac AppliChem
n-Hexane ≥97%, CHROMASOLV™ for LC-MS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Acetonitrile ≥99.9%, CHROMASOLV™, gradient grade for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
n-Octane ≥98%
Supplier: Thermo Fisher Scientific