tert-butyl (2S)-2-(aminomethyl)-5,5-difluoropiperidine-1-carboxylate 1 * 1 g
Catalog # PHBLPBLJ1884-1G
Supplier: Pharmablock (USA)
tert-butyl (2S)-2-(aminomethyl)-5,5-difluoropiperidine-1-carboxylate 1 * 1 g
Catalog # PHBLPBLJ1884-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Cyclohexane ≥99.5%, PESTINORM® for pesticide residue analysis
Supplier: VWR Chemicals
Benzyl alcohol ≥99.0% analytical reagent
Supplier: VWR Chemicals
4-Methyl-2-pentanone ≥99.0%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
(±)-Tetrahydrofurfuryl alcohol, Sigma-Aldrich®
Supplier: Merck
Tetrahydrofuran ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
Glycerine ≥97%, TECHNICAL
Supplier: VWR Chemicals
2-Propanol ≥99.5% (by GC), EMPLURA®, Supelco®
Supplier: Merck
Tetrahydrofuran ≥99.9% unstabilised, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Tetrachloroethylene ≥99.9%, SPECTRONORM® for spectroscopy
Supplier: VWR Chemicals
N,N-Dimethylformamide, extra dry over Molecular Sieve 99.8%, AcroSeal®
Supplier: Thermo Fisher Scientific
Water, CHROMASOLV™ for LC-MS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Dimethyl sulphoxide, extra dry 99.7+%, AcroSeal®
Supplier: Thermo Fisher Scientific
Acetone, J.T.Baker®
Supplier: Avantor