1-(3,3-difluoro-4-iodo-2,3-dihydro-1H-indol-1-yl)ethan-1-one 1 * 1 g
Catalog # PHBLPBLJ18210-1G
Supplier: Pharmablock (USA)
1-(3,3-difluoro-4-iodo-2,3-dihydro-1H-indol-1-yl)ethan-1-one 1 * 1 g
Catalog # PHBLPBLJ18210-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Benzyl alcohol, EMSURE® for analysis, Supelco®
Supplier: Merck
n-Pentane ≥99.5% (GC, on anhydrous basis), PESTINORM® Supra Trace
Supplier: VWR Chemicals
2-Propanol ≥70% (v/v), SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
ACETONITRILE EMPLURA® 1 * 4 L
Supplier: Merck
n-Heptane ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Cyclohexane 99+% ACS, meets analytical specifications of Reagent Ph. Eur. for spectroscopy
Supplier: Thermo Fisher Scientific
Acetonitrile, anhydrous (max. 0.001% H₂O) ≥99.95% for DNA synthesis
Supplier: VWR Chemicals
Dimethyl sulphoxide ≥99.5%, Reagent Grade
Supplier: Honeywell Chemicals
1-Butanol ≥98.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Ethanol absolute ≥99.9% (by GC), SPECTRONORM® for spectroscopy
Supplier: VWR Chemicals
n-Hexane ≥97%, CHROMASOLV™ for LC-MS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Ethyl Acetate, 99.5% for synthesis 1 * 5 L
Supplier: PanReac AppliChem
Dichloromethane ≥99.5% stabilised ACS
Supplier: Thermo Fisher Scientific
Diisopropyl ether ≥98.5% (by GC) stabilised, Puriss. p.a., Riedel-de Haën™
Supplier: Honeywell Chemicals