1-[(3,3-difluorocyclobutyl)methyl]-4-(tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole 1 * 1 g
Catalog # PHBLPBLJ0660-1G
Supplier: Pharmablock (USA)
1-[(3,3-difluorocyclobutyl)methyl]-4-(tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole 1 * 1 g
Catalog # PHBLPBLJ0660-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Ethylene glycol ≥99.7%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Acetone ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Methanol ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Xylene (mixture of isomers) ≥98.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Xylene (mixture of isomers) ≥98%, TECHNICAL
Supplier: VWR Chemicals
Water, LiChrosolv® for chromatography, for LC-MS, Supelco®
Supplier: Merck
n-Hexane ≥97%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals