ethyl (1S,3S,4S)-4-{[(tert-butoxy)carbonyl]amino}-3-hydroxycyclohexane-1-carboxylate 1 * 1 g
Catalog # PHBLPBLJ0040-1G
Supplier: Pharmablock (USA)
ethyl (1S,3S,4S)-4-{[(tert-butoxy)carbonyl]amino}-3-hydroxycyclohexane-1-carboxylate 1 * 1 g
Catalog # PHBLPBLJ0040-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
2,5-Di-tert-butylhydroquinone for synthesis, Sigma-Aldrich®
Supplier: Merck
tert-Butyl methyl ether anhydrous, 99.8 % 1 * 1 L
Supplier: Merck
5L Distilled Water 1 * 5 L
Supplier: REAGENT
Methanol ≥99.9% (by GC) Reag. Ph. Eur., ACS, ISO for analysis
Supplier: PanReac AppliChem
n-Hexane 95+%, extra pure
Supplier: Thermo Fisher Scientific
Chloroform 99.0-99.4% (by GC) stabilised, Puriss. p.a. ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
TERT. BUTANOL FOR HPLC 99,5% 1 * 1 L
Supplier: Merck
Acetonitrile 99.9%, ECD tested for residue analysis
Supplier: Thermo Fisher Scientific
Dibutyl ether, extra dry 99+%, AcroSeal®
Supplier: Thermo Fisher Scientific
Acetone ≥99.5% (by GC), liquid USP, BP, Ph. Eur., pure
Supplier: PanReac AppliChem