5-chloro-3-iodo-1-{[2-(trimethylsilyl)ethoxy]methyl}-1H-pyrazolo[4,3-b]pyridine 1 * 1 g
PHBLPBLJ00130-1G
5-chloro-3-iodo-1-{[2-(trimethylsilyl)ethoxy]methyl}-1H-pyrazolo[4,3-b]pyridine 1 * 1 g
PHBLPBLJ00130-1G
Frequently Bought Together
Toluene, extra dry over Molecular Sieve 99.85%, AcroSeal®
: Thermo Fisher Scientific
8 components: 100mg/l each of Al Cr Cu Fe Mn Ba Zn Zr in HNO3 5% 1 * 100 mL
: CUSTOM MADE CHEMICALS LAB
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
Ethanol absolute ≥99.8% (by GC), Puriss. p.a., Riedel-de Haën™
: Honeywell Chemicals
Isobutyronitrile 99%
: Thermo Fisher Scientific
Methanol ≥99.8%, Ultrapure for HPLC
: Thermo Fisher Scientific
2-Propanol 70 % pure 1 * 1 L
: PanReac AppliChem
1-Butanol ≥99.5% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
: Honeywell Chemicals
Benzonitrile 99+% for spectroscopy
: Thermo Fisher Scientific
Cyclohexane 99.5%, extra pure
: Thermo Fisher Scientific
1,5-Diphenylcarbazide, Supelco®
: Merck
Ethanol, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
N,N-Dimethylacetamide 99%, pure
: Thermo Fisher Scientific