cis-4-(3-amino-2,2,4,4-tetramethyl-cyclobutoxy)-2-chloro-benzonitrile hydrochloride 1 * 1 g
Catalog # PHBLPBLH474-1-1G
Supplier: Pharmablock (USA)
cis-4-(3-amino-2,2,4,4-tetramethyl-cyclobutoxy)-2-chloro-benzonitrile hydrochloride 1 * 1 g
Catalog # PHBLPBLH474-1-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Ethyl acetate ≥99.7%, PESTINORM® for pesticide residue analysis
Supplier: VWR Chemicals
Methanol with 0,1% (v/v) acetic acid, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
2-Methylpentane ≥99%
Supplier: Thermo Fisher Scientific
n-Octane, extra dry 99+%, AcroSeal®
Supplier: Thermo Fisher Scientific
METHANOL SUITABLE FOR HPLC >99.9% 1 * 2,5 L
Supplier: Merck
Bis(2-methoxyethyl)ether, extra dry over Molecular Sieve 99+%, AcroSeal®
Supplier: Thermo Fisher Scientific
Bromoform 99+% stabilised
Supplier: Thermo Fisher Scientific
tert-Butanol ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
ETHANOL ABSOLUTE BAKER ANALYZED 1 * 25 L
Supplier: Avantor
Benzonitrile 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
Petroleum Ether 40-60C for pesticide analysis 1 * 2,5 L
Supplier: PanReac AppliChem
Butanone (Methylethylketone) pure 1 * 2,5 L
Supplier: PanReac AppliChem
5L Distilled Water 1 * 5 L
Supplier: REAGENT