tert-butyl 4-(4-iodo-1H-pyrazol-1-yl)piperidine-1-carboxylate 1 * 5 g
PHBLPBLH1266-5G
tert-butyl 4-(4-iodo-1H-pyrazol-1-yl)piperidine-1-carboxylate 1 * 5 g
PHBLPBLH1266-5G
Frequently Bought Together
n-Hexane ≥95%, TECHNICAL
: VWR Chemicals
2-Propanol ≥99.0%, GPR RECTAPUR®
: VWR Chemicals
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
Hexane (mixture of isomers), TECHNICAL
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Ethanol 96% ≥96% (v/v), TechniSolv®, pure
: VWR Chemicals
2-Propanol ≥98%, TECHNICAL
: VWR Chemicals
Ethanol, GPR RECTAPUR®
: VWR Chemicals
1,2-Dichloroethane ≥99.8%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Ethanol absolute ≥99.5%, TechniSolv®, pure
: VWR Chemicals