3-fluoro-3-methylcyclobutan-1-amine hydrochloride 1 * 1 g
Catalog # PHBLPBLG8009-1G
Supplier: Pharmablock (USA)
3-fluoro-3-methylcyclobutan-1-amine hydrochloride 1 * 1 g
Catalog # PHBLPBLG8009-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Ethanol absolute, EMPROVE® EXPERT Ph. Eur., BP, ChP, JP, USP suitable for use as excipient, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Benzyl benzoate for synthesis, Sigma-Aldrich®
Supplier: Merck
Water, HiPerSolv CHROMANORM® for UHPLC-MS
Supplier: VWR Chemicals
Acetonitrile ≥99%, TECHNICAL
Supplier: VWR Chemicals
2-Butoxyethanol 99%, extra pure
Supplier: Thermo Fisher Scientific
tert-Butyl methyl ether, extra dry 99+%, AcroSeal®
Supplier: Thermo Fisher Scientific
Water, pure, demineralised
Supplier: Thermo Fisher Scientific
Dimethyl sulphoxide 99.7%, pure
Supplier: Thermo Fisher Scientific
Bromoform 96% stabilised
Supplier: Thermo Fisher Scientific
tert-Butanol ≥99%
Supplier: Thermo Fisher Scientific
n-Nonane 99%, Reagent Grade
Supplier: Honeywell Chemicals
N,N-Dimethylformamide, Sigma-Aldrich®
Supplier: Merck
Ethanol absolute Ph. Eur., J.T.Baker®
Supplier: Avantor
Chloroform ≥99.5% stabilised
Supplier: Honeywell Chemicals
Cyclohexane for HPLC
Supplier: Thermo Fisher Scientific