tert-butyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)cyclohex-3-ene-1-carboxylate 1 * 1 g
Catalog # PHBLPBLG122-1G
Supplier: Pharmablock (USA)
tert-butyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)cyclohex-3-ene-1-carboxylate 1 * 1 g
Catalog # PHBLPBLG122-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Ethyl acetate anhydrous, 99.8 % 1 * 2 L
Supplier: Merck
1,3-Dimethyl-2-imidazolidinone ≥99.5%, PESTINORM® for headspace gas chromatography
Supplier: VWR Chemicals
Benzyl benzoate for synthesis, Sigma-Aldrich®
Supplier: Merck
Pyridine, EMPLURA®, Supelco®
Supplier: Merck
2,5-Di-tert-butylhydroquinone for synthesis, Sigma-Aldrich®
Supplier: Merck
ACETONITRILE EMPLURA® 1 * 4 L
Supplier: Merck
Nitromethane 96% for spectroscopy
Supplier: Thermo Fisher Scientific
Water, extra pure, Deionized
Supplier: Thermo Fisher Scientific
Acetone, for GC Plus, Burdick & Jackson™
Supplier: Honeywell Chemicals
Acetonitrile ≥99.5% ACS
Supplier: Thermo Fisher Scientific
Methanol ≥99.8% ACS, meets analytical specifications of Reagent Ph. Eur.
Supplier: Thermo Fisher Scientific
tert-Butyl methyl ether ≥99.0%, AnalaR NORMAPUR® ACS
Supplier: VWR Chemicals
n-Hexane ≥97%, HiPerSolv CHROMANORM® for preparative HPLC
Supplier: VWR Chemicals
Ethanol, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
4-(Methylamino)phenol sulphate 99%, extra pure
Supplier: Thermo Fisher Scientific
Cyclopentane 95+%, pure
Supplier: Thermo Fisher Scientific