benzyl (2R)-2-(cyanomethyl)piperazine-1-carboxylate 1 * 1 g
PHBLPBJL097-1G
Frequently Bought Together
Xylene (mixture of isomers) ≥98%, TECHNICAL
: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
Methanol ≥98.5%, TECHNICAL
: VWR Chemicals
Ethanol 96% ≥96% (v/v), TechniSolv®, pure
: VWR Chemicals
2-Propanol ≥98%, TECHNICAL
: VWR Chemicals
Toluene ≥98%, TECHNICAL
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals