methyl cis-3-(hydroxymethyl)cyclohexanecarboxylate 1 * 1 g
PHBLPBJL037-1G
Frequently Bought Together
Acetonitrile, anhydrous (max. 0.001% H₂O) ≥99.8%
: VWR Chemicals
Ethyl acetate ≥99.5% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
: Honeywell Chemicals
ETHYL ACETATE 99.5% FOR SYNTHESIS 1 * 200 L
: PanReac AppliChem
Ethanol absolute ≥99.8% (by GC), Puriss. p.a., Riedel-de Haën™
: Honeywell Chemicals
Petroleum Ether 40-60C for pesticide analysis 1 * 2,5 L
: PanReac AppliChem
Toluene (Reag. Ph. Eur.) for analysis, ACS, ISO 1 * 2,5 L
: PanReac AppliChem
Benzonitrile 99+% for spectroscopy
: Thermo Fisher Scientific
Isoamyl Alcohol according to Gerber for analysis 1 * 1.000 mL
: PanReac AppliChem
Cyclohexane ≥99.7%, CHROMASOLV™ for HPLC, Riedel-de Haën™
: Honeywell Chemicals
Acetonitrile, anhydrous ≥99.8%
: Thermo Fisher Scientific
Ethanol absolute, extra dry 99.5%, AcroSeal®
: Thermo Fisher Scientific
Acetone ≥99.5%, Laboratory Reagent
: Honeywell Chemicals