(1R)-1-(2-chloro-4-fluoro-phenyl)ethanamine hydrochloride 1 * 1 g
Catalog # PHBLPBHA875-1-1G
Supplier: Pharmablock (USA)
(1R)-1-(2-chloro-4-fluoro-phenyl)ethanamine hydrochloride 1 * 1 g
Catalog # PHBLPBHA875-1-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Diisodecyl phthalate, Sigma-Aldrich®
Supplier: Merck
2-Propanol ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Methanol, anhydrous (max. 0.002% H₂O) ≥99.9%
Supplier: VWR Chemicals
Glycerine 87% 86%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
2-Propanol ≥99.0%, GPR RECTAPUR®
Supplier: VWR Chemicals
Water, AnalaR NORMAPUR® ISO 3696 Grade 3 analytical reagent
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Dimethyl sulphoxide, Uvasol® for spectroscopy, Supelco®
Supplier: Merck
Chloroform ≥99.5% stabilised
Supplier: Honeywell Chemicals
Pyridine 99+%, extra pure
Supplier: Thermo Fisher Scientific
Diethylene glycol monobutyl ether ≥99%
Supplier: Thermo Fisher Scientific
Dimethyl sulphoxide 99.7%, pure
Supplier: Thermo Fisher Scientific