trans-2-(4-ethoxy-3-fluoro-phenyl)cyclopropanamine hydrochloride 1 * 1 g
Catalog # PHBLPBGJ3222-1-1G
Supplier: Pharmablock (USA)
trans-2-(4-ethoxy-3-fluoro-phenyl)cyclopropanamine hydrochloride 1 * 1 g
Catalog # PHBLPBGJ3222-1-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Ethanol absolute (untaxed) ≥99.8% (by GC), Puriss., Riedel-de Haën™
Supplier: Honeywell Chemicals
Ethanol absolute for histology, J.T.Baker®
Supplier: Avantor
Toluene 99.8+% for analysis
Supplier: Thermo Fisher Scientific
11 components: Ca 2mg/l Zr 5mg/l K 5mg/l Zn 5mg/l Na 5mg/l Fe 5mg/l Al 5mg/l Ti 5mg/l Si 5mg/l P 10mg/l Mn 5mg/l in HNO3 5% 1 * 1 L
Supplier: CUSTOM MADE CHEMICALS LAB
2-Butoxyethanol 99%, extra pure
Supplier: Thermo Fisher Scientific
4-Methyl-2-pentanone ≥97%, TECHNICAL
Supplier: VWR Chemicals
Diisopropyl ether 98+% stabilised, pure
Supplier: Thermo Fisher Scientific
Water for analysis
Supplier: Thermo Fisher Scientific
Tetrahydrofuran, anhydrous ≥99.8% unstabilised
Supplier: Thermo Fisher Scientific
Water ACS, Deionized
Supplier: Thermo Fisher Scientific
Super-DHB ≥99.0% matrix substance for MALDI, Supelco®
Supplier: Merck
Methyl ethyl ketone 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
Methanol with 0,1% (v/v) trifluoroacetic acid, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Cyclohexane ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
N,N-Dimethylacetamide ≥99.9%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals