(1-isopropyl-3-(trifluoromethyl)-1H-pyrazol-5-yl)boronic acid 1 * 1 g
Catalog # PHBLPBEB10341-1G
Supplier: Pharmablock (USA)
(1-isopropyl-3-(trifluoromethyl)-1H-pyrazol-5-yl)boronic acid 1 * 1 g
Catalog # PHBLPBEB10341-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Ethylene glycol ≥99.7%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
n-Hexane ≥99.0% (by GC), LiChrosolv®, hypergrade for LC-MS
Supplier: Merck
n-Hexane, Uvasol® for spectroscopy, Supelco®
Supplier: Merck
Dichloromethane ≥99.9% stabilised, PESTINORM® for MOSH/MOAH analysis
Supplier: VWR Chemicals
Acetonitrile, extra dry 99.9+%, AcroSeal®
Supplier: Thermo Fisher Scientific
1,2-Dichloroethane 99+%, pure
Supplier: Thermo Fisher Scientific
Dimethyl sulphoxide ≥99%
Supplier: Thermo Fisher Scientific
Sinapic acid, Supelco®
Supplier: Merck
Ethyl acetate, extra dry over Molecular Sieve 99.9%, AcroSeal®
Supplier: Thermo Fisher Scientific
Ethanol absolute (untaxed) ≥99.8% (by GC), Puriss., Riedel-de Haën™
Supplier: Honeywell Chemicals
Trichloroethylene 99.6% stabilised ACS
Supplier: Thermo Fisher Scientific
N-Methyl-2-pyrrolidone (NMP), anhydrous ≥99.5%
Supplier: Thermo Fisher Scientific
Solvent S-316 for oil extraction
Supplier: HORIBA
n-Pentane, anhydrous 99+%, extra pure
Supplier: Thermo Fisher Scientific