2,5-dichloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine 1 * 1 g
Catalog # PHBLPBEB0734-1G
Supplier: Pharmablock (USA)
2,5-dichloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine 1 * 1 g
Catalog # PHBLPBEB0734-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Benzyl alcohol ≥99.0% analytical reagent
Supplier: VWR Chemicals
Cyclohexane, Uvasol® for spectroscopy, Supelco®
Supplier: Merck
Methanol, anhydrous (max. 0.002% H₂O) ≥99.9%
Supplier: VWR Chemicals
Methanol ≥99.5% NF, Ph. Eur., BP
Supplier: VWR Chemicals
Turpentine oil
Supplier: Roth Carl
Acetone ≥99.5% (by GC), liquid USP, BP, Ph. Eur., pure
Supplier: PanReac AppliChem
Dichloromethane, extra dry over Molecular Sieve 99.8% stabilised, AcroSeal®
Supplier: Thermo Fisher Scientific
2-Ethoxyethanol ≥99%
Supplier: Thermo Fisher Scientific
Petroleum Ether 40-60C for pesticide analysis 1 * 2,5 L
Supplier: PanReac AppliChem
2,2,4-Trimethylpentane ≥99% (by GC), CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™
Supplier: Honeywell Chemicals
n-Pentane, anhydrous ≥99.8%, over molecular sieves
Supplier: Thermo Fisher Scientific
Ethanol (taxed) Pure, meets analytical specification of USP (200 Proof), Riedel-de Haën™
Supplier: Honeywell Chemicals
Acetonitrile, anhydrous ≥99.8%, Sigma-Aldrich®
Supplier: Merck
Pyridine, extra dry over Molecular Sieve 99.5%, AcroSeal®
Supplier: Thermo Fisher Scientific