1-[3-[(1R)-1-aminoethyl]-2-fluoro-phenyl]-1,1-difluoro-2-methyl-propan-2-ol hydrochloride 1 * 1 g
Catalog # PHBLPBAH667-1-1G
Supplier: Pharmablock (USA)
1-[3-[(1R)-1-aminoethyl]-2-fluoro-phenyl]-1,1-difluoro-2-methyl-propan-2-ol hydrochloride 1 * 1 g
Catalog # PHBLPBAH667-1-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Toluene, EMPLURA®, extra pure, Supelco®
Supplier: Merck
Cyclohexane ≥99.5%, PESTINORM® for pesticide residue analysis
Supplier: VWR Chemicals
N,N-Dimethylformamide 99.0-100.00%, PESTINORM® for headspace gas chromatography
Supplier: VWR Chemicals
Acetonitrile ≥99.9% (by GC), CHROMASOLV™ for HPLC, for UV spectrophotometry, Riedel-de Haën™
Supplier: Honeywell Chemicals
1,2-Dichloroethane, extra dry over Molecular Sieve 99.5%, AcroSeal®
Supplier: Thermo Fisher Scientific
Glycerine, Sigma-Aldrich®
Supplier: Merck
Chloroform
Supplier: Roth Carl
Methanol ≥99.7% (by GC), Puriss., meets analytical specification of Ph.Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Phenetole ≥98%
Supplier: Thermo Fisher Scientific
2-Propanol 70 % pure 1 * 1 L
Supplier: PanReac AppliChem
Ethyl acetate 99.5% for analysis
Supplier: Thermo Fisher Scientific
2-Propanol 99.6% ACS, meets analytical specifications of Reagent Ph. Eur.
Supplier: Thermo Fisher Scientific
Petroleum spirit, 35…60 °C ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals