methyl (2S)-2-[[(2S)-2-[[(2S)-2-(tert-butoxycarbonylamino)-4-phenyl-butanoyl]amino]-4-methyl-pentanoyl]amino]-3-phenyl-propanoate 1 * 1 g
Catalog # PHBLPBA0625-1G
Supplier: Pharmablock (USA)
methyl (2S)-2-[[(2S)-2-[[(2S)-2-(tert-butoxycarbonylamino)-4-phenyl-butanoyl]amino]-4-methyl-pentanoyl]amino]-3-phenyl-propanoate 1 * 1 g
Catalog # PHBLPBA0625-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Ethylenediamine 99%
Supplier: Thermo Fisher Scientific
Dimethyl sulphoxide ≥99.9% USP, Multi-Compendial, J.T.Baker®
Supplier: Avantor
Dimethyl sulphoxide, anhydrous ≥99.8%
Supplier: Thermo Fisher Scientific
1-Butylboronic acid, Supelco®
Supplier: Merck
Methyl formate 98% for spectroscopy
Supplier: Thermo Fisher Scientific
Ethyl acetate ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetonitrile 99.9%, ECD tested for residue analysis
Supplier: Thermo Fisher Scientific
Water ACS
Supplier: Thermo Fisher Scientific
2,2,4-Trimethylpentane ≥99% (by GC), CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™
Supplier: Honeywell Chemicals
Cesium chloride 99.99% (trace metals basis), Optical Grade for spectroscopy
Supplier: Thermo Fisher Scientific
2-Propanol, extra dry 99.8%, AcroSeal®
Supplier: Thermo Fisher Scientific
Ethyl acetate, extra dry over Molecular Sieve 99.9%, AcroSeal®
Supplier: Thermo Fisher Scientific
Hexane (mixture of isomers) ≥95%, Laboratory Reagent
Supplier: Honeywell Chemicals
N-Ethyldiisopropylamine, Sigma-Aldrich®
Supplier: Merck
Methanol ≥99.8% for LC-MS
Supplier: Thermo Fisher Scientific