1-tert-butyl 2-methyl (2S,4S)-4-[(tert-butyldimethylsilyl)oxy]pyrrolidine-1,2-dicarboxylate 1 * 1 g
Catalog # PHBLPB98138-1G
Supplier: Pharmablock (USA)
1-tert-butyl 2-methyl (2S,4S)-4-[(tert-butyldimethylsilyl)oxy]pyrrolidine-1,2-dicarboxylate 1 * 1 g
Catalog # PHBLPB98138-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
2-Propanol ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Supplier: VWR Chemicals
Water, TECHNICAL, demineralised
Supplier: VWR Chemicals
Ethanol 96% vol Ph. Eur.
Supplier: VWR Chemicals
Dichloromethane ≥99.8% stabilised, PESTINORM® for pesticide residue analysis
Supplier: VWR Chemicals
n-Hexane ≥97%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Pyridine ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Dichloromethane ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
2-Propanol ≥98%, TECHNICAL
Supplier: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals