tert-butyl (2S)-2-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-3,6-dihydro-2H-pyridine-1-carboxylate 1 * 1 g
Catalog # PHBLPB96909-1G
Supplier: Pharmablock (USA)
tert-butyl (2S)-2-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-3,6-dihydro-2H-pyridine-1-carboxylate 1 * 1 g
Catalog # PHBLPB96909-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Methanol ≥98.5%, TECHNICAL
Supplier: VWR Chemicals
Glycerine ≥99.5%, AnalaR NORMAPUR® ACS analytical reagent, redistilled
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Ethyl acetate ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Ethanol absolute, EMPARTA® ACS analytical reagent, Supelco®
Supplier: Merck
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Supplier: VWR Chemicals
Dichloromethane stabilised, EMPLURA®
Supplier: Merck
Toluene, anhydrous (max. 0.005% H₂O) ≥99.8%, AnalaR NORMAPUR® for synthesis
Supplier: VWR Chemicals
n-Hexane ≥99%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Chloroform ≥99.9% (GC, on anhydrous basis) stabilised, PESTINORM® Supra Trace
Supplier: VWR Chemicals
Ethanol absolute ≥99.5%, TechniSolv®, pure
Supplier: VWR Chemicals
Methyl ethyl ketone ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
2-Propanol ≥99.9%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals