[(2R,3S,4R,5R)-5-(4-amino-2-oxo-pyrimidin-1-yl)-3,4-dihydroxy-tetrahydrofuran-2-yl]methyl dihydrogen phosphate 1 * 5 g
Catalog # PHBLPB95892-5G
Supplier: Pharmablock (USA)
[(2R,3S,4R,5R)-5-(4-amino-2-oxo-pyrimidin-1-yl)-3,4-dihydroxy-tetrahydrofuran-2-yl]methyl dihydrogen phosphate 1 * 5 g
Catalog # PHBLPB95892-5G
Supplier: Pharmablock (USA)
Frequently Bought Together
2-Aminophenol for synthesis, Sigma-Aldrich®
Supplier: Merck
2-Propanol ≥70% (v/v), SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Dimethyl sulphoxide, dried ≥99.9%, Puriss. p.a. (≤0.02% H₂O), Riedel-de Haën™
Supplier: Honeywell Chemicals
2-ETHOXYETHANOL, REAGENTPLUS, 99%. 1 * 1 Each
Supplier: Merck
Cesium chloride 99.99% (trace metals basis), Optical Grade for spectroscopy
Supplier: Thermo Fisher Scientific
Toluene ≥99.5% ACS
Supplier: Thermo Fisher Scientific
Benzonitrile 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
Diethyl ether ≥99% stabilised for HPLC
Supplier: Thermo Fisher Scientific
1-Chlorobutane ≥99%
Supplier: Thermo Fisher Scientific
2,2,4-Trimethylpentane 99.5% for analysis
Supplier: Thermo Fisher Scientific
Ethanolamine ACS
Supplier: Thermo Fisher Scientific
Diisopropyl ether ≥98.5% (by GC) stabilised, Puriss. p.a., Riedel-de Haën™
Supplier: Honeywell Chemicals