1-(3,5-dichloro-4-fluoro-phenyl)-2,2,2-trifluoro-ethanone 1 * 10 g
Catalog # PHBLPB95839-10G
Supplier: Pharmablock (USA)
1-(3,5-dichloro-4-fluoro-phenyl)-2,2,2-trifluoro-ethanone 1 * 10 g
Catalog # PHBLPB95839-10G
Supplier: Pharmablock (USA)
Frequently Bought Together
N,N-Dimethylformamide 99+%, extra pure
Supplier: Thermo Fisher Scientific
Dimethyl sulphoxide ≥99.7%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Dichloromethane ≥99.9% (GC, on anhydrous basis) stabilised for peptide synthesis
Supplier: VWR Chemicals
Acetone ≥99.5% (by GC), liquid USP, BP, Ph. Eur., pure
Supplier: PanReac AppliChem
N,N-Dimethylformamide ≥99.8% (by GC), Puriss. p.a. ACS, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
2-Propanol ≥99.8% (by GC), CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™
Supplier: Honeywell Chemicals
1-Pentanol 99%, pure
Supplier: Thermo Fisher Scientific
Polyvinyl alcohol, M.W. ∼ 13,000 98%, hydrolysed
Supplier: Thermo Fisher Scientific
Ethylenediamine, Sigma-Aldrich®
Supplier: Merck
Dichloromethane 99.9% stabilised, AcroSeal® for biochemistry
Supplier: Thermo Fisher Scientific
Xylene (mixture of isomers), J.T.Baker®
Supplier: Avantor
Diisopropyl ether 98+% stabilised, pure
Supplier: Thermo Fisher Scientific
(±)-2-Butanol ≥99%
Supplier: Thermo Fisher Scientific
1-Pentanol 99+% ACS
Supplier: Thermo Fisher Scientific