[(1S,2S)-2-amino-1,2-diphenyl-ethyl]-(p-tolylsulfonyl)azanide 1-isopropyl-4-methyl-benzene ruthenium(2+) chloride 1 * 1 g
Catalog # PHBLPB95483-1G
Supplier: Pharmablock (USA)
[(1S,2S)-2-amino-1,2-diphenyl-ethyl]-(p-tolylsulfonyl)azanide 1-isopropyl-4-methyl-benzene ruthenium(2+) chloride 1 * 1 g
Catalog # PHBLPB95483-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
tert-Butyl methyl ether, LiChrosolv® for HPLC, Supelco®
Supplier: Merck
Isopropyl acetate for synthesis, Sigma-Aldrich®
Supplier: Merck
Xylene (mixture of isomers) ≥98.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Chloroform 99.0-99.6% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
Hydrogen peroxide 35%, J.T.Baker®
Supplier: Avantor
Acetonitrile, LiChrosolv®, hypergrade for LC-MS, Supelco®
Supplier: Merck
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Toluene ≥99.90%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
n-Heptane, EMPLURA®, Supelco®
Supplier: Merck
Glycerine ≥97%, TECHNICAL
Supplier: VWR Chemicals
Dichloromethane ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetone ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Ethanol, GPR RECTAPUR®
Supplier: VWR Chemicals
Methanol, anhydrous (max. 0.003% H₂O) analytical reagent
Supplier: Merck
n-Hexane ≥97%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Water USP, JP, Ph. Eur., purified
Supplier: VWR Chemicals
2,4,6-Tris(2-pyridyl)-1,3,5-triazine, Supelco®
Supplier: Merck
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals