ditert-butyl-[2-(2,4,6-triisopropylphenyl)phenyl]phosphane 1 * 5 g
Catalog # PHBLPB95282-5G
Supplier: Pharmablock (USA)
ditert-butyl-[2-(2,4,6-triisopropylphenyl)phenyl]phosphane 1 * 5 g
Catalog # PHBLPB95282-5G
Supplier: Pharmablock (USA)
Frequently Bought Together
Methylcyclohexane for synthesis, Sigma-Aldrich®
Supplier: Merck
n-Hexane ≥98%, SupraSolv® for GC-MS
Supplier: Merck
Methanol, EMPARTA® ACS analytical reagent, Supelco®
Supplier: Merck
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
POTASSIUM NITRATE-15N, 98 ATOM % 15N 1 * 1 items
Supplier: Merck
N-Methyl-2-pyrrolidone (NMP) 99+% ACS
Supplier: Thermo Fisher Scientific
Ethanol absolute ≥99.8% (by GC), Puriss. p.a., Riedel-de Haën™
Supplier: Honeywell Chemicals
(±)-2-Butanol 99+% for analysis
Supplier: Thermo Fisher Scientific
Methanol, BAKER ANALYZED® for pesticide residue analysis, J.T.Baker®
Supplier: Avantor
Methanol bio-based ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Ethanolamine, Sigma-Aldrich®
Supplier: Merck
Cyclohexane, extra dry 99.5%, AcroSeal®
Supplier: Thermo Fisher Scientific