isopropyl (2R)-2-[(R)-[(2,3,4,5,6-pentafluorophenoxy)-phenoxy-phosphoryl]amino]propanoate 1 * 1 g
Catalog # PHBLPB94683-1G
Supplier: Pharmablock (USA)
isopropyl (2R)-2-[(R)-[(2,3,4,5,6-pentafluorophenoxy)-phenoxy-phosphoryl]amino]propanoate 1 * 1 g
Catalog # PHBLPB94683-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Cyclohexanone, EMPLURA®, extra pure, Supelco®
Supplier: Merck
tert-Butyl methyl ether ≥99.0% (by GC), EMPLURA®, Supelco®
Supplier: Merck
METHANOL SUITABLE FOR HPLC >99.9% 1 * 2,5 L
Supplier: Merck
Xylene - Mixture of isomers technical grade 1 * 5 L
Supplier: PanReac AppliChem
1,4-Dioxane ≥99% for HPLC
Supplier: Thermo Fisher Scientific
Water with 0,1% (v/v) trifluoroacetic acid, CHROMASOLV™ LC-MS LabReady™, Solvent Blend for LC-MS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Polyvinyl alcohol, M.W. ∼ 13,000 98%, hydrolysed
Supplier: Thermo Fisher Scientific
5L Distilled Water 1 * 5 L
Supplier: REAGENT
2-Propanol (USP, BP, Ph. Eur.) pure, pharma grade 1 * 200 L
Supplier: PanReac AppliChem
Aeroflow tap, na, general spare part 1 * 1 items
Supplier: Avantor