3-[3,5-bis(trifluoromethyl)anilino]-4-[[(R)-(6-methoxy-4-quinolyl)-[(1S,2R,4S,5R)-5-vinylquinuclidin-2-yl]methyl]amino]cyclobut-3-en e-1,2-dione 1 * 1 g
PHBLPB93648-1G
3-[3,5-bis(trifluoromethyl)anilino]-4-[[(R)-(6-methoxy-4-quinolyl)-[(1S,2R,4S,5R)-5-vinylquinuclidin-2-yl]methyl]amino]cyclobut-3-en e-1,2-dione 1 * 1 g
PHBLPB93648-1G
Frequently Bought Together
2-Methylbutane ≥99%, GPR RECTAPUR®
: VWR Chemicals
Ethanol 96% (v/v), EMPROVE® EXPERT Ph. Eur., BP, ChP, SAFC®
: MERCK PRODUCTION CHEMICALS
N-Ethyldiisopropylamine ≥99.5% for peptide synthesis
: VWR Chemicals
Water, CHROMASOLV™ for LC-MS, Riedel-de Haën™
: Honeywell Chemicals
Xylene (mixture of isomers) ≥98%, liquid
: PanReac AppliChem
N,N-Dimethylacetamide 99.8+% for HPLC
: Thermo Fisher Scientific
1,4-Dioxane ≥99% for HPLC
: Thermo Fisher Scientific
Acrylonitrile 99+%
: Thermo Fisher Scientific
Water, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
: Honeywell Chemicals
2-Propanol ≥99.9%, HiPerSolv CHROMANORM® for LC-MS
: VWR Chemicals
Bis(2-methoxyethyl)ether, extra dry 99+%, AcroSeal®
: Thermo Fisher Scientific
ACETONE HPLC 2.5L 1 * 2,5 L
: LAB 3