chromic 2,4-ditert-butyl-6-[[[(1S,2S)-2-[(3,5-ditert-butyl-6-oxido-cyclohexa-2,4-dien-1-ylidene)methylamino]cyclohexyl]amino]methyle ne]cyclohexa-2,4-dien-1-olate 1 * 1 g
Catalog # PHBLPB92616-1G
Supplier: Pharmablock (USA)
chromic 2,4-ditert-butyl-6-[[[(1S,2S)-2-[(3,5-ditert-butyl-6-oxido-cyclohexa-2,4-dien-1-ylidene)methylamino]cyclohexyl]amino]methyle ne]cyclohexa-2,4-dien-1-olate 1 * 1 g
Catalog # PHBLPB92616-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Cyclohexanone ≥99.0% analytical reagent
Supplier: VWR Chemicals
n-Hexane, LiChrosolv® for liquid chromatography, Supelco®
Supplier: Merck
Acetonitrile, LiChrosolv®, hypergrade for LC-MS, Supelco®
Supplier: Merck
Toluene ≥99.90%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Xylene (mixture of isomers) ≥98.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Water, AnalaR NORMAPUR® ISO 3696 Grade 3 analytical reagent
Supplier: VWR Chemicals
Pyridine ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Diethyl ether stabilised, TECHNICAL, rectified
Supplier: VWR Chemicals
Tetrahydrofuran ≥99.9% unstabilised, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Methyl ethyl ketone ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Acetone ≥99.8%, SupraSolv® for GC-ECD/FID, Supelco®
Supplier: Merck
Ethanol 96% vol Ph. Eur.
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Ethyl acetate ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals