(3R)-N-{3,6-dichloro-1H-pyrazolo[3,4-d]pyrimidin-4-yl}piperidin-3-amine hydrochloride 1 * 1 g
PHBLPB92508-1-1G
(3R)-N-{3,6-dichloro-1H-pyrazolo[3,4-d]pyrimidin-4-yl}piperidin-3-amine hydrochloride 1 * 1 g
PHBLPB92508-1-1G
Frequently Bought Together
Ethylenediamine 99%
: Thermo Fisher Scientific
N-Methyl-2-pyrrolidone (NMP) 99.5% for analysis
: Thermo Fisher Scientific
Acetone, for GC Plus, Burdick & Jackson™
: Honeywell Chemicals
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
2,2,4-Trimethylpentane 99.5% for analysis
: Thermo Fisher Scientific
1,2-Dichlorobenzene 99+% for HPLC
: Thermo Fisher Scientific
Acetonitrile 99.9%, ECD tested for residue analysis
: Thermo Fisher Scientific
Dibutyl ether, extra dry 99+%, AcroSeal®
: Thermo Fisher Scientific
N-Methyl-2-pyrrolidone (NMP) ≥99.0% ACS, Riedel-de Haën™
: Honeywell Chemicals
tert-Butyl methyl ether ≥99.0%, AnalaR NORMAPUR® ACS
: VWR Chemicals
Xylene (mixture of isomers) ≥98%, liquid
: PanReac AppliChem
Acetonitrile, anhydrous ≥99.8%
: Thermo Fisher Scientific