(1R)-2,3-dihydro-1H-inden-1-amine hydrochloride 1 * 5 g
Catalog # PHBLPB90788-1-5G
Supplier: Pharmablock (USA)
(1R)-2,3-dihydro-1H-inden-1-amine hydrochloride 1 * 5 g
Catalog # PHBLPB90788-1-5G
Supplier: Pharmablock (USA)
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Glycerine 85% Ph. Eur.
Supplier: VWR Chemicals
Glycerine ≥98%, GPR RECTAPUR®
Supplier: VWR Chemicals
Ethanol 96% (v/v), EMPROVE® EXPERT Ph. Eur., BP, ChP, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
2-Propanol, EMPROVE® ESSENTIAL Ph. Eur., BP, JP, USP, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
2,5-Di-tert-butylhydroquinone for synthesis, Sigma-Aldrich®
Supplier: Merck
Cyclohexanone ≥99.0% analytical reagent
Supplier: VWR Chemicals
Water, extra pure, Deionized
Supplier: Thermo Fisher Scientific
n-Heptane, anhydrous 99+%
Supplier: Thermo Fisher Scientific
Tetrahydrofuran ≥99.0% contains inhibitor, Reagent Grade
Supplier: Honeywell Chemicals
Methanol ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Methanol ≥99.8% ACS, meets analytical specifications of Reagent Ph. Eur.
Supplier: Thermo Fisher Scientific