O1-tert-butyl O2-methyl (2R,4R)-4-fluoropyrrolidine-1,2-dicarboxylate 1 * 1 g
Catalog # PHBLPB06278-1G
Supplier: Pharmablock (USA)
O1-tert-butyl O2-methyl (2R,4R)-4-fluoropyrrolidine-1,2-dicarboxylate 1 * 1 g
Catalog # PHBLPB06278-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Supplier: VWR Chemicals
Glycerine ≥97%, TECHNICAL
Supplier: VWR Chemicals
n-Octane 97%, pure
Supplier: Thermo Fisher Scientific
Acetone ≥99.5% (by GC), liquid USP, BP, Ph. Eur., pure
Supplier: PanReac AppliChem
1-Octanol ≥99%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
N,N-Dimethylacetamide ≥99%
Supplier: Thermo Fisher Scientific
Petroleum Ether 40-60C for pesticide analysis 1 * 2,5 L
Supplier: PanReac AppliChem
Toluene 99.85% for HPLC
Supplier: Thermo Fisher Scientific
2-Ethoxyethanol ≥99%
Supplier: Thermo Fisher Scientific
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Dimethyl sulphoxide ≥99.5%, Reagent Grade
Supplier: Honeywell Chemicals
Dichloromethane, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
Tetrahydrofuran ≥99.9% stabilised, Puriss. p.a. ACS, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Methanol 99.8% for HPLC
Supplier: Thermo Fisher Scientific
Acetone ≥99.5%, Laboratory Reagent
Supplier: Honeywell Chemicals