4-(tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole 1 * 25 g
Catalog # PHBLPB06068-25G
Supplier: Pharmablock (USA)
4-(tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole 1 * 25 g
Catalog # PHBLPB06068-25G
Supplier: Pharmablock (USA)
Frequently Bought Together
Ethyl acetate ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
1-Propanol ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Chloroform 99.0-99.6% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
Water, SupraSolv® for gas chromatography, Supelco®
Supplier: Merck
Ethanolamine ≥99%, GPR RECTAPUR® for liquid scintillation counting
Supplier: VWR Chemicals
Ethanol, GPR RECTAPUR®
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Supplier: VWR Chemicals
2,2,4-Trimethylpentane ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
n-Hexane, dehydrated (max. 0.01% H₂O) ≥95%, GPR RECTAPUR® for synthesis
Supplier: VWR Chemicals
Acetone ≥99.8% (by GC), CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™
Supplier: Honeywell Chemicals
Cyclohexane 99.8% for HPLC
Supplier: Thermo Fisher Scientific
N,N-Dimethylformamide, Sigma-Aldrich®
Supplier: Merck
Petroleum Ether 40-60C for pesticide analysis 1 * 2,5 L
Supplier: PanReac AppliChem
Acetone ≥99.5%, Laboratory Reagent
Supplier: Honeywell Chemicals
Pyridine, extra dry over Molecular Sieve 99.5%, AcroSeal®
Supplier: Thermo Fisher Scientific
Acetonitrile 99.8% for HPLC
Supplier: Thermo Fisher Scientific
Methanol, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
Tetrahydrofuran, extra dry 99.85% unstabilised, AcroSeal®
Supplier: Thermo Fisher Scientific