7-azabicyclo[2.2.1]heptane-2-carboxylic acid ethyl ester hydrochloride 1 * 1 g
Catalog # PHBLPB05739-1-1G
Supplier: Pharmablock (USA)
7-azabicyclo[2.2.1]heptane-2-carboxylic acid ethyl ester hydrochloride 1 * 1 g
Catalog # PHBLPB05739-1-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
N-HEPTANE FOR LIQUID CHROMATOGRAPHY 1 * 185 L
Supplier: Merck
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Methanol, LiChrosolv® for liquid chromatography, Supelco®
Supplier: Merck
Methanol ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
2-Propanol 70% ≥70% (v/v), GPR RECTAPUR®
Supplier: VWR Chemicals
Isobutanol ≥99%
Supplier: Thermo Fisher Scientific
Chloroform ≥99.0% stabilised BP, USP, NF for UV spectrophotometry, for IR spectrophotometry, for HPLC
Supplier: PanReac AppliChem
o-Xylene, extra dry 99%, AcroSeal®
Supplier: Thermo Fisher Scientific
Toluene 99.8+% for analysis
Supplier: Thermo Fisher Scientific
Benzyl alcohol ≥99.9% (by GC), CHROMASOLV™ for headspace gas chromatography, Riedel-de Haën™
Supplier: Honeywell Chemicals
Acetonitrile, extra dry over Molecular Sieve 99.9%, AcroSeal®
Supplier: Thermo Fisher Scientific
n-Hexane, extra dry 96+%, AcroSeal®
Supplier: Thermo Fisher Scientific
Diethyl ether, anhydrous 99+% stabilised ACS, meets analytical specifications of Reagent Ph. Eur.
Supplier: Thermo Fisher Scientific