(2S,3R,4R,5R)-2,4-bis(benzoyloxy)-5-[(benzoyloxy)methyl]-3-methyloxolan-3-yl benzoate 1 * 5 g
Catalog # PHBLPB00556-5G
Supplier: Pharmablock (USA)
(2S,3R,4R,5R)-2,4-bis(benzoyloxy)-5-[(benzoyloxy)methyl]-3-methyloxolan-3-yl benzoate 1 * 5 g
Catalog # PHBLPB00556-5G
Supplier: Pharmablock (USA)
Frequently Bought Together
Dimethyl sulphoxide, EMSURE® ACS for analysis, Supelco®
Supplier: Merck
(±)-Tetrahydrofurfuryl alcohol, Sigma-Aldrich®
Supplier: Merck
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Methanol ≥99.9% (by GC) Reag. Ph. Eur., ACS, ISO for analysis
Supplier: PanReac AppliChem
Chloroform 99.0-99.4% (by GC) stabilised, Puriss. p.a. ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Diisopropyl ether ≥98.5% (by GC) stabilised, Puriss. p.a., Riedel-de Haën™
Supplier: Honeywell Chemicals
1,2-Dimethoxyethane, extra dry 99+% stabilised, AcroSeal®
Supplier: Thermo Fisher Scientific
Ethanolamine ACS
Supplier: Thermo Fisher Scientific
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
n-Hexane 95%, ChromAR®, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
5L Distilled Water 1 * 5 L
Supplier: REAGENT