3-methoxyazetidine hydrochloride 1 * 10 g
Catalog # PHBLPB00226-01-10G
Supplier: Pharmablock (USA)
Frequently Bought Together
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for preparative HPLC
Supplier: VWR Chemicals
2-Propanol ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Toluene ≥98%, TECHNICAL
Supplier: VWR Chemicals
Ethanol denatured 95% ≥95%, GPR RECTAPUR® (denatured with methanol)
Supplier: VWR Chemicals
Aniline for synthesis, Sigma-Aldrich®
Supplier: Merck
Diethyl ether ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
n-Heptane, LiChrosolv® for liquid chromatography, Supelco®
Supplier: Merck
1,3-Dioxolane ≥99.0% stabilised, GPR RECTAPUR® for synthesis
Supplier: VWR Chemicals
Acrylonitrile 99+%
Supplier: Thermo Fisher Scientific
Ethanol (untaxed), anhydrous, tested according to Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
N,N-Dimethylacetamide ≥99%
Supplier: Thermo Fisher Scientific
Chloroform ≥99.8% stabilised ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Ethyl acetate 99+%, extra pure
Supplier: Thermo Fisher Scientific
Acetonitrile, extra dry over Molecular Sieve 99.9%, AcroSeal®
Supplier: Thermo Fisher Scientific
Toluene ≥99.7% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals