azetidin-3-one hydrochloride 1 * 10 g
PHBLPB00225-1-10G
Frequently Bought Together
Diethyl ether ≥99% stabilised, GPR RECTAPUR®
: VWR Chemicals
Glycerine 85% Ph. Eur.
: VWR Chemicals
Formamide ≥99.0%, AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals