(3S)-tetrahydrofuran-3-amine hydrochloride 1 * 5 g
PHBLPB00222-1-5G
Frequently Bought Together
n-Hexane ≥97%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Hexane (mixture of isomers), TECHNICAL
: VWR Chemicals
n-Heptane ≥99.0%, AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
Xylene (mixture of isomers) ≥98%, TECHNICAL
: VWR Chemicals
Ethanol absolute ≥99.5%, TechniSolv®, pure
: VWR Chemicals
Diethyl ether ≥99.5% stabilised, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Acetone ≥99.8%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals