Order Entry
United Kingdom
ContactUsLinkComponent
PhaseTwo D5 Dewar 5 litre Static Holding Time (days) - 13 Normal Evaporation Rate (liters/day) - 0.4 Liquid Nitrogen Capacity - 5 litres Neck Diameter (mm) - 36 Overall Height / Diameter (mm) - 390 x 234 Weight Empty / Full (kg) - 3 / 7 (Warranty is 1 * 1 items
Catalog # TAYW10-152-01-0000
Supplier:  TAYLOR WHARTON
PhaseTwo D5 Dewar 5 litre Static Holding Time (days) - 13 Normal Evaporation Rate (liters/day) - 0.4 Liquid Nitrogen Capacity - 5 litres Neck Diameter (mm) - 36 Overall Height / Diameter (mm) - 390 x 234 Weight Empty / Full (kg) - 3 / 7 (Warranty is 1 * 1 items
Catalog # TAYW10-152-01-0000
Supplier:  TAYLOR WHARTON
Frequently Bought Together