(3R,4S,5R,6R)-3,4,5-TRIS((TRIMETHYLSILYL)OXY)-6-(((TRIMETHYLSILYL)OXY)METHYL)TETRAHYDRO-2H-PYRAN-2-ONE 1 * 100 g
Catalog # MSPPM23043-100G
Supplier: MARKET SOURCE PART PROCESS
(3R,4S,5R,6R)-3,4,5-TRIS((TRIMETHYLSILYL)OXY)-6-(((TRIMETHYLSILYL)OXY)METHYL)TETRAHYDRO-2H-PYRAN-2-ONE 1 * 100 g
Catalog # MSPPM23043-100G
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together
4-(Methylamino)phenol sulphate 99%
Supplier: Thermo Fisher Scientific
Acetonitrile ≥99.5% ACS
Supplier: Thermo Fisher Scientific
Chlorobenzene ≥99%
Supplier: Thermo Fisher Scientific
Chloroform 99.0-99.4% (by GC) stabilised, Puriss. p.a. ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
(±)-2-Methyltetrahydrofuran 99+% stabilised, pure
Supplier: Thermo Fisher Scientific
Dimethyl Sulfoxide (USP, BP, Ph. Eur.) pharma grade 1 * 190 L
Supplier: PanReac AppliChem
n-Hexane 95%, ChromAR®, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
Petroleum spirit, 30…40 °C, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Mineral oil light, white NF, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
Toluene ≥99.7% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
tert-Butyl methyl ether ≥99.0%, AnalaR NORMAPUR® ACS
Supplier: VWR Chemicals
N,N-Dimethylacetamide 99%, pure
Supplier: Thermo Fisher Scientific
Methanol ≥99.9%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals