Order Entry
United Kingdom
ContactUsLinkComponent
Minitron 1 x 25 mm throw on 120mm base for floor mounting Without cooling Temperature Range – ambient -+5°C to 65°C Speed Range 20 – 400 rpm Power Supply 230V 50Hz Pre-drilled tray for use with clamps Clamp for 16 x 50ml Falcon Tubes 1 * 1 items
Catalog # INFOAN130586
Supplier:  INFORS
Minitron 1 x 25 mm throw on 120mm base for floor mounting Without cooling Temperature Range – ambient -+5°C to 65°C Speed Range 20 – 400 rpm Power Supply 230V 50Hz Pre-drilled tray for use with clamps Clamp for 16 x 50ml Falcon Tubes 1 * 1 items
Catalog # INFOAN130586
Supplier:  INFORS
Frequently Bought Together