Order Entry
United Kingdom
ContactUsLinkComponent
XBridge BEH HILIC Column, 130Å, 5 µm, 2.1 mm X 100 mm 1 * 1 items
Catalog # WATE186004445
Supplier:  WATERS
XBridge BEH HILIC Column, 130Å, 5 µm, 2.1 mm X 100 mm 1 * 1 items
Catalog # WATE186004445
Supplier:  WATERS
Frequently Bought Together