Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
2-Propanol ≥98%, TECHNICAL

2-Propanol ≥98%, TECHNICAL

950 L
Catalog # 20922.930
Flammable
Irritant
Supplier:   VWR Chemicals
2-Propanol ≥98%, TECHNICAL
Catalog # 20922.930
Supplier:  VWR Chemicals
CAS Number:  67-63-0
2-Propanol ≥98%, TECHNICAL
2-Propanol ≥98%, TECHNICAL

950 L

CAS Number:  67-63-0
Price
Inquire For Price
Log in for availability
1 * 950 L
Product Details & Documents
Synonyms:  Isopropyl rubbing alcohol, Isopropanol, 2-Hydroxy propane, IPA, Dimethyl carbinol, Isopropyl alcohol, Propan-2-ol
  • CAS Number:67-63-0
  • Formula:(CH₃)₂CHOH
  • MDL Number:MFCD00011674
  • Molecular Weight:60.1 g/mol
  • Merck Index:13,05228
  • Boiling Pt:82 °C (1013 hPa)
  • Melting Pt:–89 °C
  • Density:0.786 g/cm³ (20 °C)
  • Flash Pt:12 °C
  • Storage Temperature:Ambient
  • EINECS:200-661-7
  • UN:1219
  • ADR:3,II
  • REACH:01-2119457558-25
Specifications

Specification Test Results

  • Assay:
    Min. 98 %

Specifications

  • Pk:
    950 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.